Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XID | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336434.1 | NP_001324212.1 | 34 - 35 | 4816 | 113 | |
NM_001336435.1 | NP_001324208.1 | 34 - 35 | 4362 | 113 | ||
NM_001336436.1 | NP_001324211.1 | 34 - 35 | 4383 | 113 | ||
NM_001336437.1 | NP_001324213.1 | 20 - 21 | 2873 | 113 | ||
NM_001336438.1 | NP_001324210.1 | 34 - 35 | 4644 | 113 | ||
NM_001336439.1 | NP_001324209.1 | 35 - 36 | 4338 | 113 | ||
NM_001336440.1 | NP_001324214.1 | 35 - 36 | 4338 | 113 | ||
NM_128883.3 | NP_180882.2 | 34 - 35 | 4401 | 113 | ||
GenBank mRNA | LC177513.1 | - | 33 - 34 | 4329 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |