Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128940.2 | NP_565771.1 | 1 - 2 | 638 | 64 | |
GenBank mRNA | AJ011633.1 | - | 1 - 2 | 321 | 64 | |
AK118179.1 | - | 1 - 2 | 415 | 64 | ||
BT005443.1 | - | 1 - 2 | 297 | 64 | ||
Y09427.1 | - | 321 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |