Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RRP41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125406.1 | NP_001118878.1 | 3 - 4 | 392 | 67 | |
NM_116027.5 | NP_191721.1 | 2 - 3 | 234 | 67 | ||
GenBank mRNA | AF191741.1 | - | 2 - 3 | 123 | 67 | |
AK118846.1 | - | 2 - 3 | 150 | 67 | ||
BT005485.1 | - | 1 - 2 | 52 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |