Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARFA1E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203232.1 | NP_001190161.1 | 4 - 5 | 547 | 103 | |
NM_001203233.1 | NP_001190162.1 | 4 - 5 | 547 | 103 | ||
NM_116094.4 | NP_191788.1 | 4 - 5 | 460 | 103 | ||
GenBank mRNA | AF372902.1 | - | 4 - 5 | 439 | 103 | |
AY057728.1 | - | 3 - 4 | 332 | 103 | ||
AY085382.1 | - | 4 - 5 | 439 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |