Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UNE10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340226.1 | NP_001329601.1 | 2 - 3 | 818 | 124 | |
NM_001340227.1 | NP_001329602.1 | 3 - 4 | 743 | 124 | ||
NM_116222.4 | NP_191916.3 | 3 - 4 | 888 | 124 | ||
GenBank mRNA | AK117229.1 | - | 3 - 4 | 735 | 124 | |
BT025971.1 | - | 3 - 4 | 664 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |