Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YAB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084868.2 | NP_001078337.1 | 2 - 3 | 853 | 100 | |
NM_116235.3 | NP_567154.1 | 3 - 4 | 937 | 100 | ||
GenBank mRNA | AF136540.1 | - | 3 - 4 | 558 | 100 | |
AK316696.1 | - | 2 - 3 | 448 | 100 | ||
AY037186.1 | - | 3 - 4 | 742 | 100 | ||
BT002662.1 | - | 3 - 4 | 450 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |