Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ROPGEF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125431.2 | NP_001118903.1 | 3 - 4 | 685 | 70 | |
NM_001340253.1 | NP_001328183.1 | 2 - 3 | 613 | 70 | ||
NM_116271.3 | NP_191956.2 | 3 - 4 | 685 | 70 | ||
GenBank mRNA | BT030385.1 | - | 2 - 3 | 422 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |