Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATMYC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160722.2 | NP_001154194.1 | 2 - 3 | 540 | 61 | |
NM_001340255.1 | NP_001329692.1 | 2 - 3 | 576 | 61 | ||
NM_116272.4 | NP_191957.2 | 2 - 3 | 685 | 61 | ||
GenBank mRNA | AB493664.1 | - | 2 - 3 | 443 | 61 | |
AF251697.1 | - | 2 - 3 | 443 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |