Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340311.1 | NP_001329076.1 | 4 - 5 | 331 | 75 | |
NM_116372.3 | NP_192051.2 | 4 - 5 | 331 | 75 | ||
GenBank mRNA | AF192885.2 | - | 4 - 5 | 328 | 75 | |
AF290435.1 | - | 4 - 5 | 328 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |