Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SWN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340357.1 | NP_001328435.1 | 15 - 16 | 2394 | 72 | |
NM_116433.3 | NP_567221.1 | 15 - 16 | 2414 | 72 | ||
GenBank mRNA | AF100163.1 | - | 15 - 16 | 2368 | 72 | |
AY057477.1 | - | 15 - 16 | 2394 | 72 | ||
AY090293.1 | - | 15 - 16 | 2301 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |