Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SULTR3;2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116504.3 | NP_192179.1 | 3 - 4 | 782 | 70 | |
GenBank mRNA | AB004060.1 | - | 3 - 4 | 742 | 70 | |
AK229084.1 | - | 3 - 4 | 782 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |