Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATG8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340494.1 | NP_001329707.1 | 2 - 3 | 755 | 78 | |
NM_116700.2 | NP_192371.1 | 3 - 4 | 569 | 78 | ||
NM_178967.4 | NP_849298.1 | 3 - 4 | 639 | 78 | ||
GenBank mRNA | AB073176.1 | - | 2 - 3 | 148 | 78 | |
AK227679.1 | - | 3 - 4 | 185 | 78 | ||
AY084859.1 | - | 3 - 4 | 280 | 78 | ||
BT006385.1 | - | 2 - 3 | 91 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |