Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104479.2 | NP_175997.1 | 1 - 2 | 330 | 100 | |
GenBank mRNA | AF198054.1 | - | 1 - 2 | 281 | 100 | |
AY085996.1 | - | 1 - 2 | 290 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Arabidopsis thaliana Probe spans exons |