Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333869.1 | NP_001321983.1 | 1 - 2 | 176 | 70 | |
NM_104677.5 | NP_176193.2 | 1 - 2 | 223 | 70 | ||
NM_202322.2 | NP_974051.1 | 1 - 2 | 223 | 70 | ||
GenBank mRNA | AK175262.1 | - | 1 - 2 | 123 | 70 | |
AK175368.1 | - | 1 - 2 | 123 | 70 | ||
AK221539.1 | - | 1 - 2 | 123 | 70 | ||
BT010443.1 | - | 1 - 2 | 116 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |