Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AAE15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117482.4 | NP_193143.2 | 6 - 7 | 1068 | 72 | |
GenBank mRNA | AY062804.1 | - | 6 - 7 | 1037 | 72 | |
BT010332.1 | - | 6 - 7 | 919 | 72 | ||
EF093797.1 | - | 6 - 7 | 919 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |