Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AAE15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117482.4 | NP_193143.2 | 9 - 10 | 1389 | 106 | |
GenBank mRNA | AY062804.1 | - | 9 - 10 | 1358 | 106 | |
BT010332.1 | - | 9 - 10 | 1240 | 106 | ||
EF093797.1 | - | 9 - 10 | 1240 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |