Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEE48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117483.4 | NP_193144.1 | 1 - 2 | 108 | 68 | |
GenBank mRNA | AY099580.1 | - | 1 - 2 | 108 | 68 | |
BT002150.1 | - | 1 - 2 | 42 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |