Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117488.4 | NP_193147.1 | 4 - 5 | 566 | 99 | |
GenBank mRNA | AF395067.1 | - | 4 - 5 | 445 | 99 | |
L32874.1 | - | 4 - 5 | 498 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |