Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APRR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084936.2 | NP_001078405.1 | 8 - 9 | 1574 | 59 | |
NM_001160778.2 | NP_001154250.1 | 8 - 9 | 1610 | 59 | ||
NM_001203830.1 | NP_001190759.1 | 8 - 9 | 1597 | 59 | ||
NM_001341252.1 | NP_001329342.1 | 9 - 10 | 1588 | 59 | ||
NM_001341253.1 | NP_001329343.1 | 9 - 10 | 1568 | 59 | ||
NM_001341254.1 | NP_001329339.1 | 8 - 9 | 1849 | 59 | ||
NM_001341255.1 | NP_001329340.1 | 8 - 9 | 1829 | 59 | ||
NM_001341256.1 | NP_001329341.1 | 8 - 9 | 1428 | 59 | ||
NM_117912.4 | NP_567548.1 | 8 - 9 | 1610 | 59 | ||
NM_179072.3 | NP_849403.1 | 8 - 9 | 1582 | 59 | ||
NM_179073.4 | NP_849404.2 | 8 - 9 | 1550 | 59 | ||
GenBank mRNA | AB041531.1 | - | 7 - 8 | 1300 | 59 | |
AF385725.1 | - | 8 - 9 | 1434 | 59 | ||
AY143963.1 | - | 7 - 8 | 1290 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |