Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341273.1 | NP_001329801.1 | 3 - 4 | 1170 | 85 | |
NM_117939.5 | NP_193563.3 | 3 - 4 | 546 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |