Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_117980.5 | NP_193599.3 | 2 - 3 | 800 | 67 | |
GenBank mRNA | FJ708745.1 | - | 1 - 2 | 85 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |