Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334433.1 | NP_001323064.1 | 16 - 17 | 3605 | 60 | |
NM_105674.6 | NP_177163.3 | 17 - 18 | 3438 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |