Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105828.4 | NP_177315.1 | 2 - 3 | 1050 | 88 | |
GenBank mRNA | AK229787.1 | - | 2 - 3 | 688 | 88 | |
AK230407.1 | - | 2 - 3 | 687 | 88 | ||
BT008311.1 | - | 2 - 3 | 481 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |