Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_123052.5 | NP_198510.2 | 1 - 2 | 337 | 83 | |
GenBank mRNA | AK117747.1 | - | 1 - 2 | 142 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |