Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GORK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344207.1 | NP_001332118.1 | 10 - 11 | 2612 | 66 | |
NM_001344208.1 | NP_001332117.1 | 9 - 10 | 2324 | 66 | ||
NM_001344209.1 | NP_001332119.1 | 9 - 10 | 2688 | 66 | ||
NM_123109.5 | NP_198566.2 | 11 - 12 | 2546 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |