Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCPL43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036274.1 | NP_001031351.1 | 5 - 6 | 676 | 88 | |
NM_001335380.1 | NP_001323740.1 | 5 - 6 | 799 | 88 | ||
NM_001335381.1 | NP_001323739.1 | 5 - 6 | 799 | 88 | ||
NM_126893.4 | NP_178937.2 | 5 - 6 | 676 | 88 | ||
GenBank mRNA | BT004306.1 | - | 5 - 6 | 676 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |