Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH6B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084418.1 | NP_001077887.1 | 13 - 14 | 1274 | 64 | |
NM_001084419.1 | NP_001077888.1 | 14 - 15 | 1474 | 64 | ||
NM_126989.2 | NP_179032.1 | 14 - 15 | 1474 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |