Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100266.3 | NP_171882.1 | 5 - 6 | 902 | 69 | |
NM_202026.2 | NP_973755.1 | 4 - 5 | 860 | 69 | ||
NM_202027.2 | NP_973756.1 | 5 - 6 | 905 | 69 | ||
GenBank mRNA | AF370317.1 | - | 5 - 6 | 771 | 69 | |
AY063102.1 | - | 4 - 5 | 669 | 69 | ||
U66592.1 | - | 5 - 6 | 763 | 69 | ||
U89791.1 | - | 5 - 6 | 752 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |