Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS15A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331728.1 | NP_001318947.1 | 4 - 5 | 553 | 64 | |
NM_100651.5 | NP_172256.1 | 3 - 4 | 489 | 64 | ||
NM_202054.3 | NP_973783.1 | 3 - 4 | 417 | 64 |
Gene Symbol: AT5G59850 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF001412.1 | - | 3 - 4 | 371 | 64 | |
AF332447.1 | - | 2 - 3 | 295 | 64 | ||
AF360284.1 | - | 3 - 4 | 384 | 64 | ||
AF386922.1 | - | 3 - 4 | 378 | 64 | ||
AK317713.1 | - | 3 - 4 | 411 | 64 | ||
AY042830.1 | - | 3 - 4 | 383 | 64 | ||
AY051040.1 | - | 2 - 3 | 295 | 64 | ||
AY081472.1 | - | 2 - 3 | 295 | 64 | ||
AY081689.1 | - | 2 - 3 | 295 | 64 | ||
AY088335.1 | - | 3 - 4 | 389 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |