Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LTL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111300.4 | NP_187079.1 | 3 - 4 | 784 | 83 | |
GenBank mRNA | AY085128.1 | - | 3 - 4 | 677 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |