Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111477.4 | NP_187254.1 | 1 - 2 | 452 | 72 | |
GenBank mRNA | AB000799.1 | - | 1 - 2 | 340 | 72 | |
AY069917.1 | - | 1 - 2 | 434 | 72 | ||
BT001024.1 | - | 1 - 2 | 299 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |