Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KOB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111689.5 | NP_187467.1 | 2 - 3 | 1026 | 93 | |
GenBank mRNA | AF543710.1 | - | 2 - 3 | 548 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |