Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WRKY20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341825.1 | NP_001329239.1 | 3 - 4 | 1026 | 70 | |
NM_118798.2 | NP_567752.1 | 2 - 3 | 886 | 70 | ||
NM_179119.5 | NP_849450.1 | 2 - 3 | 690 | 70 | ||
GenBank mRNA | AF425837.1 | - | 2 - 3 | 387 | 70 | |
AJ630479.1 | - | 2 - 3 | 387 | 70 | ||
AY150436.1 | - | 1 - 2 | 171 | 70 | ||
AY568651.1 | - | 2 - 3 | 387 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |