Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VIIIB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001341848.1 | NP_001328308.1 | 4 - 5 | 813 | 137 | |
NM_001341849.1 | NP_001328309.1 | 3 - 4 | 752 | 137 | ||
NM_001341850.1 | NP_001328307.1 | 4 - 5 | 829 | 137 | ||
NM_118871.6 | NP_194467.5 | 3 - 4 | 620 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |