Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAMS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334857.1 | NP_001321745.1 | 1 - 2 | 556 | 99 | |
NM_001334858.1 | NP_001321744.1 | 1 - 2 | 556 | 99 | ||
NM_001334859.1 | NP_001321746.1 | 1 - 2 | 556 | 99 | ||
NM_001334860.1 | NP_001321747.1 | 1 - 2 | 556 | 99 | ||
NM_148667.2 | NP_683508.1 | 2 - 3 | 389 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |