Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBP13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202935.1 | NP_001189864.1 | 28 - 29 | 3303 | 147 | |
NM_001337961.1 | NP_001319526.1 | 28 - 29 | 3303 | 147 | ||
NM_001337962.1 | NP_001326292.1 | 28 - 29 | 3306 | 147 | ||
NM_112024.6 | NP_187797.3 | 28 - 29 | 3232 | 147 | ||
GenBank mRNA | BT002760.1 | - | 28 - 29 | 3033 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |