Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MKK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036674.2 | NP_001031751.1 | 2 - 3 | 253 | 93 | |
NM_001125607.1 | NP_001119079.1 | 2 - 3 | 219 | 93 | ||
NM_119127.4 | NP_194710.1 | 2 - 3 | 307 | 93 | ||
GenBank mRNA | AB015313.1 | - | 2 - 3 | 171 | 93 | |
AF385688.1 | - | 2 - 3 | 190 | 93 | ||
AY078009.1 | - | 2 - 3 | 164 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |