Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VLN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203940.1 | NP_001190869.1 | 20 - 21 | 2529 | 106 | |
NM_001341999.1 | NP_001329918.1 | 20 - 21 | 2570 | 106 | ||
NM_001342000.1 | NP_001329917.1 | 19 - 20 | 2500 | 106 | ||
NM_119162.5 | NP_194745.1 | 19 - 20 | 2842 | 106 | ||
GenBank mRNA | AK117296.1 | - | 19 - 20 | 2449 | 106 | |
BT005980.1 | - | 18 - 19 | 2239 | 106 | ||
Y12782.1 | - | 2482 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |