Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119233.2 | NP_567859.1 | 8 - 9 | 1176 | 88 | |
GenBank mRNA | AY050894.1 | - | 8 - 9 | 1148 | 88 | |
AY096675.1 | - | 8 - 9 | 1074 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |