Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: G18F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345098.1 | NP_001332742.1 | 5 - 6 | 1960 | 145 | |
NM_124854.4 | NP_200284.1 | 6 - 7 | 2663 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |