Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342225.1 | NP_001328575.1 | 9 - 10 | 1984 | 82 | |
NM_119519.4 | NP_567929.1 | 9 - 10 | 2134 | 82 | ||
NM_179157.3 | NP_849488.1 | 9 - 10 | 2134 | 82 | ||
GenBank mRNA | AY059742.1 | - | 9 - 10 | 1923 | 82 | |
AY117173.1 | - | 9 - 10 | 1866 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |