Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UVH6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100201.3 | NP_171818.1 | 12 - 13 | 2288 | 84 | |
NM_179253.1 | NP_849584.1 | 11 - 12 | 2351 | 84 | ||
GenBank mRNA | AY062471.1 | - | 12 - 13 | 2288 | 84 | |
AY090788.1 | - | 11 - 12 | 2351 | 84 | ||
AY090915.1 | - | 11 - 12 | 2151 | 84 | ||
BT010344.1 | - | 11 - 12 | 2151 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |