Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAMMA-ADAPTIN 1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332624.1 | NP_001322140.1 | 5 - 6 | 1398 | 115 | |
NM_102238.2 | NP_173802.1 | 6 - 7 | 978 | 115 | ||
NM_179370.4 | NP_849701.1 | 5 - 6 | 1135 | 115 | ||
GenBank mRNA | BT004307.1 | - | 5 - 6 | 1051 | 115 | |
BT005557.1 | - | 5 - 6 | 835 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |