Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sks11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112184.4 | NP_187947.1 | 2 - 3 | 1471 | 85 | |
GenBank mRNA | BT003958.1 | - | 2 - 3 | 1410 | 85 | |
BT005058.1 | - | 2 - 3 | 1364 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |