Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AME3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036697.1 | NP_001031774.1 | 9 - 10 | 939 | 128 | |
NM_001342166.1 | NP_001328058.1 | 7 - 8 | 1267 | 128 | ||
NM_119418.3 | NP_194992.1 | 8 - 9 | 1175 | 128 | ||
NM_202937.1 | NP_974666.1 | 8 - 9 | 1065 | 128 | ||
GenBank mRNA | BT029776.1 | - | 8 - 9 | 797 | 128 | |
D45355.1 | - | 8 - 9 | 856 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |