Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUC95A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119590.4 | NP_195152.2 | 5 - 6 | 1704 | 98 | |
GenBank mRNA | AY125494.1 | - | 5 - 6 | 1573 | 98 | |
BT002722.1 | - | 5 - 6 | 1569 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |