Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119600.4 | NP_567965.1 | 7 - 8 | 1173 | 89 | |
GenBank mRNA | AY042877.1 | - | 7 - 8 | 1143 | 89 | |
AY081454.1 | - | 7 - 8 | 1067 | 89 | ||
AY168881.1 | - | 7 - 8 | 1091 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |