Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THY-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342289.1 | NP_001328947.1 | 8 - 9 | 1269 | 108 | |
NM_001342290.1 | NP_001328948.1 | 7 - 8 | 1227 | 108 | ||
NM_001342291.1 | NP_001328949.1 | 8 - 9 | 1172 | 108 | ||
NM_001342292.1 | NP_001328950.1 | 7 - 8 | 1123 | 108 | ||
NM_119623.5 | NP_195183.2 | 8 - 9 | 1269 | 108 | ||
GenBank mRNA | AK227728.1 | - | 8 - 9 | 1208 | 108 | |
BT003159.1 | - | 5 - 6 | 968 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |