Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDG37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_179638.2 | NP_849969.2 | 9 - 10 | 1068 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |