Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_179685.2 | NP_850016.1 | 1 - 2 | 417 | 65 | |
GenBank mRNA | AB039925.1 | - | 1 - 2 | 278 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |